4-Iodphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Iodophenyl)boronic acid, Dihydroxy(4-iodophenyl)borane, 4-Iodobenzeneboronic acid, (4-Iodophenyl)metaboric acid

M-Clarity™ quality level = MQ200

471933-25G 471933-5G
SIAL471933-25GEA 347 EUR
SIAL471933-25G SIAL471933-5G
4-Iodphenylboronsäure, Sigma-Aldrich®
4-Iodphenylboronsäure
Formel: C₆H₆BIO₂
MDL Number: MFCD01319014
CAS-Nummer: 5122-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR