(±)-γ-Octalacton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one

M-Clarity™ quality level = MQ300

W279609-1KG-K W279609-5KG-K
SIALW279609-1KG-KEA 131 EUR
SIALW279609-1KG-K SIALW279609-5KG-K
(±)-γ-Octalacton, Sigma-Aldrich®
(±)-γ-Octalacton
Formel: C₈H₁₄O₂
Siedepunkt: 234 °C (1013 hPa)
Dichte: 0.991 g/cm³ (0 °C)
MDL Number: MFCD00005402
CAS-Nummer: 104-50-7
EINECS: 203-208-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR