(±)-γ-Octalacton ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(±)-Octan-4-olide, 5-Butyldihydrofuran-2(3H)-one
A18142.22
A18142.14
A18142.22EA
181
EUR
A18142.22
A18142.14
(±)-γ-Octalacton ≥98%
(±)-γ-Octalacton
Purity: 98%
CAS No.: 104-50-7
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 126°(259°F)
Refractive Index: 1.444
Boiling Point: 233-235°
MDL No.: MFCD00005402
Formel:
C₈H₁₄O₂ Siedepunkt: 233…235 °C Dichte: 0.981 Flammpunkt: 126 °C (259 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005402 CAS-Nummer: 104-50-7 EINECS: 203-208-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...