4-Acetylphenylboronsäure
Lieferant: Thermo Scientific
Synonyme:
(4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane
SB02098.1
SB02098.10
SB02098.50
MAYBSB02098.1EA
0
EUR
MAYBSB02098.1
MAYBSB02098.10
MAYBSB02098.50
4-Acetylphenylboronsäure
4-Acetylphenylboronsäure
Formel:
C₈H₉BO₃ Molekulargewicht: 163.97 |
MDL Number:
MFCD01074667 CAS-Nummer: 149104-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...