3-Acetylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3'-Boronoacetophenone, 3-Acetylphenylboronic Acid, 3-Acetylbenzeneboronic acid, Dihydroxy(3-acetylphenyl)borane, (3-Acetylphenyl)metaboric acid, 3-Acetylphenylboronic acid, 3-Acetylbenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

470813-25G 470813-5G
SIAL470813-25GEA 207 EUR
SIAL470813-25G SIAL470813-5G
3-Acetylphenylboronsäure, Sigma-Aldrich®
3-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molekulargewicht: 163.97
MDL Number: MFCD01074678
CAS-Nummer: 204841-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR