5-Brom-2-hydroxybenzylalkohol 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-Bromo-2-(hydroxymethyl)phenol, (5-Bromo-2-hydroxyphenyl)methanol
331480050
331480250
ACRO331480050EA
59.9
EUR
ACRO331480050
ACRO331480250
5-Brom-2-hydroxybenzylalkohol 98%
5-Brom-2-hydroxybenzylalkohol
Formel:
C₇H₇BrO₂ Siedepunkt: 302 °C (1013 hPa) Schmelzpunkt: 106…111 °C |
MDL Number:
MFCD00004618 CAS-Nummer: 2316-64-5 EINECS: 219-026-0 Merck Index: 12,01460 |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 106 to 111 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...