4-Iod-2,6-xylenol

Lieferant: Thermo Scientific

Synonyme: 2-Hydroxy-5-iodo-3-xylene, 4-Iodo-2,6-dimethylphenol, 2,6-Dimethyl-4-iodophenol

RDP00223DA RDP00223EA RDP00223EE
MAYBRDP00223DAEA 0 EUR
MAYBRDP00223DA MAYBRDP00223EA MAYBRDP00223EE
4-Iod-2,6-xylenol
4-Iod-2,6-xylenol
Formel: C₈H₉IO
MDL Number: MFCD00094415
CAS-Nummer: 10570-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR