3,5-Dibrom-p-kresol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,3-Dibromo-5-hydroxy-2-methylbenzene, 2,6-Dibromo-4-hydroxytoluene, 3,5-Dibromo-4-methylphenol
L00660.03
L00660.06
L00660.03EA
86.6
EUR
L00660.03
L00660.06
3,5-Dibrom-p-kresol ≥97%
3,5-Dibrom-p-kresol
Purity: 97%
CAS No.: 13979-81-2
Molecular Formula: C7H6Br2O
Formula Weight: 265.94
Melting Point: 106-110°
Beilstein Reference: 2249692
Formel:
C₇H₆Br₂O Schmelzpunkt: 106…110 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00013968 CAS-Nummer: 13979-81-2 EINECS: 237-763-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...