2,4-Diisopropylphenol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,3-Di(methylethyl)-4-hydroxybenzene, 2,4-Bisisopropylphenol

557811-1G 557811-5G
SIAL557811-1GEA 131 EUR
SIAL557811-1G SIAL557811-5G
2,4-Diisopropylphenol, Sigma-Aldrich®
2,4-Diisopropylphenol
Formel: C₁₂H₁₈O
MDL Number: MFCD01707536
CAS-Nummer: 2934-05-6
EINECS: 220-906-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR