Butylisovalerat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isovaleric acid butyl ester, Butyl 3-methylbutanoate

M-Clarity™ quality level = MQ300

W221805-1KG-K W221805-20KG-K W221805-4KG-K
SIALW221805-1KG-KEA 90 EUR
SIALW221805-1KG-K SIALW221805-20KG-K SIALW221805-4KG-K
Butylisovalerat, Sigma-Aldrich®
Butylisovalerat
Formel: C₉H₁₈O₂
MDL Number: MFCD00048775
CAS-Nummer: 109-19-3
EINECS: 203-654-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR