5-Fluor-3-indolylessigsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
F4506-1G
F4506-250MG
SIALF4506-1GEA
1720
EUR
SIALF4506-1G
SIALF4506-250MG
5-Fluor-3-indolylessigsäure, Sigma-Aldrich®
5-Fluor-3-indolylessigsäure
Formel:
C₁₀H₈FNO₂ |
MDL Number:
MFCD00056921 CAS-Nummer: 443-73-2 EINECS: 207-138-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...