4-Brompyridiniumchlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

M-Clarity™ quality level = MQ100

B80429-1G B80429-5G
SIALB80429-1GEA 28.2 EUR
SIALB80429-1G SIALB80429-5G
4-Brompyridiniumchlorid, Sigma-Aldrich®
4-Bromopyridinium chloride
Formel: C₅H₅BrClN
Schmelzpunkt: 235…238 °C
MDL Number: MFCD00012828
CAS-Nummer: 19524-06-2
EINECS: 243-128-4
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR