1-Ethyl-3-methylimidazoliumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Ethyl-1-methyl-1H-imidazol-3-ium bromide

M-Clarity™ quality level = MQ100

89483-50G-F 89483-5G-F
SIAL89483-50G-FEA 183 EUR
SIAL89483-50G-F SIAL89483-5G-F
1-Ethyl-3-methylimidazoliumbromid, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium bromide
Formel: C₆H₁₁BrN₂
MDL Number: MFCD03427610
CAS-Nummer: 65039-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR