1-Hexyl-3-methylimidazoliumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-n-Hexyl-3-methylimidazolium chloride
M-Clarity™ quality level = MQ100
87929-50G
87929-5G
SIAL87929-50GEA
154
EUR
SIAL87929-50G
SIAL87929-5G
1-Hexyl-3-methylimidazoliumchlorid, Sigma-Aldrich®
1-Hexyl-3-methylimidazolium chloride
Formel:
C₁₀H₁₉ClN₂ Schmelzpunkt: –85 °C Dichte: 1.05 g/cm³ (20 °C) |
MDL Number:
MFCD03093289 CAS-Nummer: 171058-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...