3,4,5-Trimethoxybenzylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

338931-25G 338931-5G
SIAL338931-25GEA 201 EUR
SIAL338931-25G SIAL338931-5G
3,4,5-Trimethoxybenzylamin, Sigma-Aldrich®
3,4,5-Trimethoxybenzylamin
Formel: C₁₀H₁₅NO₃
Siedepunkt: 106 °C (0.1 mmHg)
Dichte: 1.155 g/cm³ (20 °C)
MDL Number: MFCD00017150
CAS-Nummer: 18638-99-8
EINECS: 242-468-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR