Desoxycholsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
12α-Hydroxylithocholic acid, (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, DCA, 12alpha-Hydroxylithocholic acid, 3α,12α-Dihydroxycholan-24-oic acid, 3α,12α-Dihydroxycholanic acid
M-Clarity™ quality level = MQ100
Formel:
C₂₄H₄₀O₄ Siedepunkt: 504 °C (1013 hPa) Schmelzpunkt: 190 °C |
MDL Number:
MFCD00003673 CAS-Nummer: 83-44-3 EINECS: 201-478-5 Merck Index: 13,02916 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...