Natriumcholat Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme: 3α,7α,12α-Trihydroxycholanic acid sodium salt hydrate, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate hydrate, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt hydrate, Cholic acid sodium salt hydrate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt hydrate, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt hydrate
27029-100G-F 27029-500G-F
SIAL27029-100G-FEA 190 EUR
SIAL27029-100G-F SIAL27029-500G-F
Natriumcholat Hydrat, Sigma-Aldrich®
Natriumcholat Hydrat
Formel: C₂₄H₃₉NaO₅·H₂O
Molekulargewicht: 448.58 g/mol
MDL Number: MFCD00064138
CAS-Nummer: 206986-87-0
EINECS: 206-643-5