Tris(triphenylphosphin)rhodium(I)chlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

M-Clarity™ quality level = MQ200

199982-1G 199982-5G
SIAL199982-1GEA 194 EUR
SIAL199982-1G SIAL199982-5G
Tris(triphenylphosphin)rhodium(I)chlorid, Sigma-Aldrich®
Tris(triphenylphosphin)rhodium(I)chlorid
Formel: C₅₄H₄₅ClP₃Rh
Schmelzpunkt: 157 °C
Dichte: 1.363 g/cm³ (20 °C)
MDL Number: MFCD00010016
CAS-Nummer: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR