Dinatriumsuccinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic

M-Clarity™ quality level = MQ200

14160-100G 14160-500G
SIAL14160-100GEA 23 EUR
SIAL14160-100G SIAL14160-500G
Dinatriumsuccinat, Sigma-Aldrich®
Dinatriumsuccinat
Formel: C₄H₄Na₂O₄
Dichte: 1.88 g/cm³ (30 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002790
CAS-Nummer: 150-90-3
EINECS: 205-778-7
Merck Index: 12,08828

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR