2-Amino-5-nitrothiazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Nitrothiazol-2-ylamine, 5-Nitrothiazol-2-amine
M-Clarity™ quality level = MQ100
133507-100G
133507-25G
SIAL133507-100GEA
135
EUR
SIAL133507-100G
SIAL133507-25G
2-Amino-5-nitrothiazol, Sigma-Aldrich®
2-Amino-5-nitrothiazole
Formel:
C₃H₃N₃O₂S Schmelzpunkt: 203 °C |
MDL Number:
MFCD00005326 CAS-Nummer: 121-66-4 EINECS: 204-490-9 Merck Index: 13,00456 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...