tert-Butylethylether, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Ethoxy-2-methylpropane
M-Clarity™ quality level = MQ100
03202-25ML
03202-5ML
SIAL03202-25MLEA
236
EUR
SIAL03202-25ML
SIAL03202-5ML
tert-Butylethylether, Sigma-Aldrich®
tert-Butylethylether
Formel:
C₆H₁₄O Siedepunkt: 71 °C (1013 hPa) Schmelzpunkt: –97 °C Dichte: 0.752 g/cm³ (20 °C) |
MDL Number:
MFCD00009225 CAS-Nummer: 637-92-3 EINECS: 211-309-7 UN-Nummer: 1179 ADR: 3,II Merck Index: 13,03809 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...