Palladium(II)nitrat Dihydrat

Lieferant: Roth Carl
Gefahr

Synonyme: Palladium dinitrate dihydrate

9026.1 9026.2
ROTH9026.1EA 0 EUR
ROTH9026.1 ROTH9026.2
Palladium(II)nitrat Dihydrat
Palladium(II)nitrat Dihydrat
Formel: N₂O₆Pd·2H₂O
CAS-Nummer: 32916-07-7
EINECS: 233-265-8
UN-Nummer: 1477
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR