Trimethylsulfoxoniumiodid
Lieferant: Thermo Scientific
SB01336.1
SB01336.10
SB01336.50
MAYBSB01336.1EA
0
EUR
MAYBSB01336.1
MAYBSB01336.10
MAYBSB01336.50
Trimethylsulfoxoniumiodid
Trimethylsulphoxonium iodide
Formel:
C₃H₉IOS Schmelzpunkt: 165…175 °C |
MDL Number:
MFCD00011899 CAS-Nummer: 1774-47-6 EINECS: 217-204-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...