α,α,α-Trifluor-p-toluonitril
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluoromethyl)benzonitrile, 4-Cyanobenzotrifluoride, p-(Trifluoromethyl)benzonitrile
SB00652.1
SB00652.10
SB00652.50
MAYBSB00652.1EA
0
EUR
MAYBSB00652.1
MAYBSB00652.10
MAYBSB00652.50
α,α,α-Trifluor-p-toluonitril
α,α,α-Trifluor-p-toluonitril
Formel:
C₈H₄F₃N Siedepunkt: 183…185 °C (1013 hPa) Schmelzpunkt: 35…38 °C Dichte: 1.2784 g/cm³ (20 °C) Flammpunkt: 72 °C (closed cup) |
MDL Number:
MFCD00001826 CAS-Nummer: 455-18-5 EINECS: 207-239-1 UN-Nummer: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...