4-Isopropoxybenzaldehyd
Lieferant: Thermo Scientific
RDR03610.10
RDR03610.50
MAYBRDR03610.10EA
0
EUR
MAYBRDR03610.10
MAYBRDR03610.50
4-Isopropoxybenzaldehyd
4-Isopropoxybenzaldehyd
Formel:
C₁₀H₁₂O₂ |
MDL Number:
MFCD00052357 CAS-Nummer: 18962-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...