α-Brom-4-chlor-2-fluortoluol

Lieferant: Thermo Scientific

KM10077DA KM10077EA
MAYBKM10077DAEA 0 EUR
MAYBKM10077DA MAYBKM10077EA
α-Brom-4-chlor-2-fluortoluol
α-Brom-4-chlor-2-fluortoluol
Formel: C₇H₅BrClF
MDL Number: MFCD00831105
CAS-Nummer: 71916-82-0
UN-Nummer: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR