3-Acetyl-2,5-dichlorthiophen
Lieferant: Thermo Scientific
Synonyme:
1-(2,5-Dichloro-3-thienyl)-1-ethanone
BTB14077.1
BTB14077.10
BTB14077.50
MAYBBTB14077.1EA
0
EUR
MAYBBTB14077.1
MAYBBTB14077.10
MAYBBTB14077.50
3-Acetyl-2,5-dichlorthiophen
3-Acetyl-2,5-dichlorthiophen
Formel:
C₆H₄Cl₂OS Siedepunkt: 120…122 °C (6 mmHg) Schmelzpunkt: 37…40 °C |
MDL Number:
MFCD00014522 CAS-Nummer: 36157-40-1 EINECS: 252-893-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...