2-Iodfluoren

Lieferant: Thermo Scientific

BTB10887.1 BTB10887.10
MAYBBTB10887.1EA 0 EUR
MAYBBTB10887.1 MAYBBTB10887.10
2-Iodfluoren
2-Iodfluoren
Formel: C₁₃H₉I
MDL Number: MFCD00019049
CAS-Nummer: 2523-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR