3-Brom-7-nitro-1H-indazol

Lieferant: Cayman Chemical

Synonyme: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

81345-100 81345-50 81345-250 81345-25
CAYM81345-100EA 148 EUR
CAYM81345-100 CAYM81345-50 CAYM81345-250 CAYM81345-25
3-Brom-7-nitro-1H-indazol
3-Bromo-7-nitro-1H-indazole
Formel: C₇H₄BrN₃O₂
Schmelzpunkt: 176 °C
MDL Number: MFCD00159910
CAS-Nummer: 74209-34-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR