3,5-Dimethylbenzolsulfonylchlorid ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3,5-Dimethylbenzene-1-sulphonyl chloride, m-Xylene-5-sulphonyl chloride
L18603.03
L18603.06
L18603.03EA
37
EUR
L18603.03
L18603.06
3,5-Dimethylbenzolsulfonylchlorid ≥97%
3,5-Dimethylbenzolsulfonylchlorid
Purity: 97%
CAS No.: 2905-27-3
Molecular Formula: C8H9ClO2S
Formula Weight: 204.68
Melting Point: 92-94°
Beilstein Reference: 2640617
UN No.: 3261
MDL No.: MFCD03094655
Formel:
C₈H₉ClO₂S Schmelzpunkt: 92…94 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03094655 CAS-Nummer: 2905-27-3 UN-Nummer: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...