EDTA Dikaliumsalz Dihydrat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt
Purity: 99%
CAS No.: 25102-12-9
Molecular Formula: C10H14K2N2O8.2H2O
Formula Weight: 404.46 (368.43anhy)
Odour: Odourless
Melting Point: ca 272° dec.
MDL No.: MFCD00150036
Formel:
C₁₀H₁₄K₂N₂O₈·2H₂O Schmelzpunkt: ∼ 272 °C (dec.) Dichte: 1 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150036 CAS-Nummer: 25102-12-9 EINECS: 217-895-0 Merck Index: 14,3517 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...