N,O-Bis(trimethylsilyl)acetamid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
Purity: 95%
CAS No.: 10416-59-8
Molecular Formula: C8H21NOSi2
Formula Weight: 203.43
Storage Temperature: 2°C to 8°C
Flash Point: 42°(107°F)
Refractive Index: 1.4170
Melting Point: -24°
Boiling Point: 71-73°/35mm
Beilstein Reference: 1306669
UN No.: 2920
MDL No.: MFCD00008270
Formel:
C₈H₂₁NOSi₂ Siedepunkt: 71…73 °C (35 mmHg) Schmelzpunkt: –24 °C Dichte: 0.829 Flammpunkt: 42 °C (107 °F) |
MDL Number:
MFCD00008270 CAS-Nummer: 10416-59-8 EINECS: 233-892-7 UN-Nummer: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...