1-Aminopyridiniumiodid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B25703.06
B25703.14
B25703.06EA
55.4
EUR
B25703.06
B25703.14
1-Aminopyridiniumiodid 97%
1-Aminopyridinium iodide
Purity: 97%
CAS No.: 6295-87-0
Molecular Formula: C5H7IN2
Formula Weight: 222.03
Melting Point: ca 160° dec.
Beilstein Reference: 3713241
Formel:
C₅H₇IN₂ Schmelzpunkt: ∼ 160 °C (Zersetzung) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00051893 CAS-Nummer: 6295-87-0 EINECS: 228-566-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...