α-Brom-3-nitro-p-tolylsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B25255.03
B25255.06
B25255.14
B25255.03EA
37.1
EUR
B25255.03
B25255.06
B25255.14
α-Brom-3-nitro-p-tolylsäure ≥97%
α-Brom-3-nitro-p-tolylsäure
Purity: 97%
CAS No.: 55715-03-2
Molecular Formula: C8H6BrNO4
Formula Weight: 260.05
Melting Point: 128-133°
UN No.: 3261
MDL No.: MFCD00216575
Formel:
C₈H₆BrNO₄ Schmelzpunkt: 128…133 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00216575 CAS-Nummer: 55715-03-2 UN-Nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...