α,α,α-Trifluor-o-xylol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23453.03
B23453.06
B23453.03EA
44.7
EUR
B23453.03
B23453.06
α,α,α-Trifluor-o-xylol ≥97%
α,α,α-Trifluor-o-xylol
Purity: 97%
CAS No.: 13630-19-8
Molecular Formula: C8H7F3
Formula Weight: 160.14
Refractive Index: 1.4350
Boiling Point: 125-126°
UN No.: 1993
MDL No.: MFCD00153204
Formel:
C₈H₇F₃ Siedepunkt: 125…126 °C Dichte: 1.174 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00153204 CAS-Nummer: 13630-19-8 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...