4-Brom-α,α,α,2,3,5,6-heptafluortoluol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Bromo-2,3,5,6-tetrafluorobenzotrifluoride, 1-Bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
B21350.03
B21350.06
B21350.03EA
32.2
EUR
B21350.03
B21350.06
4-Brom-α,α,α,2,3,5,6-heptafluortoluol ≥99%
4-Brom-α,α,α,2,3,5,6-heptafluortoluol
Purity: 99%
CAS No.: 17823-46-0
Molecular Formula: C7BrF7
Formula Weight: 296.98
Boiling Point: 152-153°
MDL No.: MFCD00075240
Formel:
C₇BrF₇ Siedepunkt: 152…153 °C Dichte: 1.929 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075240 CAS-Nummer: 17823-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...