N,N-Diethyl-1,1,1-trimethylsilylamin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Trimethylsilyldiethylamine, (Diethylamino)trimethylsilane, N,N-Diethylaminotrimethylsilane, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, TMSDEA
Purity: 98%
CAS No.: 996-50-9
Molecular Formula: (CH3)3SiN(CH2CH3)2
Formula Weight: 145.32
Flash Point: 10°(50°F)
Refractive Index: 1.4110
Odour: Amine-like
Melting Point: -10°
Boiling Point: 125-126°
Beilstein Reference: 635718
UN No.: 2733
MDL No.: MFCD00009040
Formel:
C₇H₁₉NSi Siedepunkt: 125…126 °C Schmelzpunkt: –10 °C Dichte: 0.760 Flammpunkt: 10 °C (50 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009040 CAS-Nummer: 996-50-9 EINECS: 213-637-6 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...