Natriumcholat Hydrat ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
3α,7α,12α-Trihydroxycholanic acid sodium salt hydrate, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate hydrate, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt hydrate, Cholic acid sodium salt hydrate, Sodium 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoate hydrate, 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt hydrate, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt hydrate
Purity: 99%
CAS No.: 206986-87-0
Molecular Formula: C24H39NaO5.xH2O
Formula Weight: 430.56(anhy)
Beilstein Reference: 3582354
Optical Rotation: +32° (c=0.5 in water)
Formel:
C₂₄H₃₉NaO₅·H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00064138 CAS-Nummer: 206986-87-0 EINECS: 206-643-5 |