5-Methylbenzimidazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Methyl-1H-benzimidazole, 5-Methyl-1H-benzo[d]imidazole
A15957.06
A15957.14
A15957.22
A15957.06EA
44.3
EUR
A15957.06
A15957.14
A15957.22
5-Methylbenzimidazol ≥98%
5-Methylbenzimidazole
Purity: 98%
CAS No.: 614-97-1
Molecular Formula: C8H8N2
Formula Weight: 132.17
Melting Point: 112-117°
Boiling Point: 169-172°/1mm
MDL No.: MFCD00010740
Formel:
C₈H₈N₂ Siedepunkt: 169…172 °C (1 mmHg) Schmelzpunkt: 112…117 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010740 CAS-Nummer: 614-97-1 EINECS: 210-401-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...