(Methyl)triphenylphosphoniumiodid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A15644.14
A15644.22
A15644.36
A15644.14EA
65.6
EUR
A15644.14
A15644.22
A15644.36
(Methyl)triphenylphosphoniumiodid ≥98%
(Methyl)triphenylphosphoniumiodid
Purity: 98%
CAS No.: 2065-66-9
Molecular Formula: CH3P(C6H5)3I
Formula Weight: 404.23
Odour: Odourless
Melting Point: 185-188°
UN No.: 3464
MDL No.: MFCD00066175
Formel:
C₁₉H₁₈IP Schmelzpunkt: 185…188 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066175 CAS-Nummer: 2065-66-9 EINECS: 218-178-5 UN-Nummer: 3464 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...