3-(2-Methoxyphenyl)propionsäure ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
3-(2-Methoxyphenyl)propanoic acid
A14314.14
A14314.06
A14314.22
A14314.14EA
64.2
EUR
A14314.14
A14314.06
A14314.22
3-(2-Methoxyphenyl)propionsäure ≥98%
3-(2-Methoxyphenyl)propionsäure
Purity: 98+%
CAS No.: 6342-77-4
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 85-88°
Beilstein Reference: 2097012
Formel:
C₁₀H₁₂O₃ Siedepunkt: 302 °C (1013 hPa) Schmelzpunkt: 85…88 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002772 CAS-Nummer: 6342-77-4 EINECS: 228-738-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...