5-Chlor-o-anisidin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A14244.22
A14244.36
A14244.22EA
51.1
EUR
A14244.22
A14244.36
5-Chlor-o-anisidin ≥98%
5-Chlor-o-anisidin
Purity: 98%
CAS No.: 95-03-4
Molecular Formula: C7H8ClNO
Formula Weight: 157.60
Odour: Characteristic
Melting Point: 79-84°
Boiling Point: 135°/8mm
Beilstein Reference: 879161
UN No.: 2233
MDL No.: MFCD00007777
Formel:
C₇H₈ClNO Siedepunkt: 135 °C (8 mmHg) Schmelzpunkt: 79…84 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007777 CAS-Nummer: 95-03-4 EINECS: 202-385-2 UN-Nummer: 2233 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...