2-Mercapto-1-methylimidazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Mercapto-N-methyl-1H-imidazole, Thiamazole, Methimazole, 1-Methyl-1H-imidazole-2(3H)-thione, 2-Mercapto-N-methylimidazole, 2-Mercapto-1-methyl-1H-imidazole
Purity: 98%
CAS No.: 60-56-0
Molecular Formula: C4H6N2S
Formula Weight: 114.17
Odour: Stench
Melting Point: 144-147°
Boiling Point: 280° dec.
Solubility: Freely soluble in water. Soluble in alcohol, chloroform. Sparingly soluble in ether, benzene.
Beilstein Reference: 108646
Merck Reference: 14,5971
MDL No.: MFCD00179321
Formel:
C₄H₆N₂S Molekulargewicht: 114.17 g/mol Siedepunkt: 280 °C (Zers.) Schmelzpunkt: 144…147 °C Dichte: 1.36 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00179321 CAS-Nummer: 60-56-0 EINECS: 200-482-4 Merck Index: 13,06000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...