2,5-Diphenyloxazol 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
PPO, 2,5-Diphenyl-1,3-oxazole
A10654.14
A10654.22
A10654.36
A10654.14EA
39
EUR
A10654.14
A10654.22
A10654.36
2,5-Diphenyloxazol 99%
2,5-Diphenyloxazole
Purity: 99%
CAS No.: 92-71-7
Molecular Formula: C15H11NO
Formula Weight: 221.26
Melting Point: 70-74°
Boiling Point: 360°
MDL No.: MFCD00005306
Formel:
C₁₅H₁₁NO Siedepunkt: 360 °C Schmelzpunkt: 70…74 °C Dichte: 1.060 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005306 CAS-Nummer: 92-71-7 EINECS: 202-181-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...