1-(2-Bromethyl)naphthalin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

43491.06 43491.03
43491.06EA 324 EUR
43491.06 43491.03
1-(2-Bromethyl)naphthalin ≥97%
1-(2-Bromoethyl)naphthalene

Purity: 97% CAS No.: 13686-49-2 Molecular Formula: C12H11Br Formula Weight: 235.13 Flash Point: 110°(230°F) Physical Form: Liquid Boiling Point: 237-238°1.65 MDL No.: MFCD00037737

Formel: C₁₂H₁₁Br
Siedepunkt: 237…238 °C1.65
Flammpunkt: >110 °C (230 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00037737
CAS-Nummer: 13686-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR