Zinn(II)oxalat

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tin (II) ethanedioate, Oxalic acid tin (II) salt

14113.22 14113.36
14113.22EA 40.8 EUR
14113.22 14113.36
Zinn(II)oxalat
Zinn(II)oxalat

CAS No.: 814-94-8 Molecular Formula: C2O4Sn Formula Weight: 206.72 Physical Form: Powder Density: 3.56 Appearance: White Melting Point: 280° dec. Solubility: Soluble in dilute HCl. Insoluble in water Beilstein Reference: 3708588 Merck Reference: 14,8786 UN No.: 3288 MDL No.: MFCD00040678

Formel: C₂O₄Sn
Schmelzpunkt: 280 °C (dec.)
Dichte: 3.56
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00040678
CAS-Nummer: 814-94-8
EINECS: 212-414-0
UN-Nummer: 3288
ADR: 6.1,III
Merck Index: 14,8786

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR