Dysprosiumtrinitrat Pentahydrat ≥99,9%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Dysprosium (III) nitrate pentahydrate
12922.14
12922.22
12922.14EA
80.7
EUR
12922.14
12922.22
Dysprosiumtrinitrat Pentahydrat ≥99,9%
Dysprosiumtrinitrat Pentahydrat
Purity: 99.9%
CAS No.: 10031-49-9
Molecular Formula: Dy(NO3)3.5H2O
Formula Weight: 438.59 (348.51anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Cream to pale yellow
Odour: Odourless
pH: 3
Melting Point: 88.6°
Merck Reference: 14,3482
UN No.: 1477
MDL No.: MFCD00149683
Formel:
DyN₃O₉·5H₂O Molekulargewicht: 438.59 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149683 CAS-Nummer: 10031-49-9 EINECS: 233-410-5 UN-Nummer: 1477 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...