Tris(triphenylphosphin)rhodium(I)chlorid ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium
Purity: 97%
CAS No.: 14694-95-2
Molecular Formula: C54H45ClP3Rh
Formula Weight: 925.23
Storage Temperature: 2°C to 8°C
Physical Form: Micro Crystals
Odour: Odourless
Melting Point: ca 250° dec.
Solubility: Soluble in most solvents (e.g. benzene, ethanol, chloroform, dichloromethane) but with phosphine dissociation. Reacts with O2 in solution
Merck Reference: 14,10047
MDL No.: MFCD00010016
Formel:
C₅₄H₄₅ClP₃Rh Schmelzpunkt: ∼ 250 °C (dec.) Dichte: 1.363 g/cm³ (20 °C) |
MDL Number:
MFCD00010016 CAS-Nummer: 14694-95-2 EINECS: 238-744-5 Merck Index: 14,10047 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...