1-Hexyl-3-methylimidazoliumhexafluorophosphat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
354180250
354180050
354181000
ACRO354180250EA
204
EUR
ACRO354180250
ACRO354180050
ACRO354181000
1-Hexyl-3-methylimidazoliumhexafluorophosphat 99%
1-Hexyl-3-methylimidazolium hexafluorophosphate
Formel:
C₁₀H₁₉F₆N₂P Schmelzpunkt: –73.5 °C Dichte: 1.3045 g/cm³ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03093296 CAS-Nummer: 304680-35-1 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4200 (20°C, 589 nm) |
NMR | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...