(Methyl)triphenylphosphoniumiodid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Methyltriphenylphosphonium iodide
224030250
224031000
ACRO224030250EA
56.1
EUR
ACRO224030250
ACRO224031000
(Methyl)triphenylphosphoniumiodid 97%
(Methyl)triphenylphosphoniumiodid
Formel:
C₁₉H₁₈IP Schmelzpunkt: 185…187 °C |
MDL Number:
MFCD00066175 CAS-Nummer: 2065-66-9 EINECS: 218-178-5 UN-Nummer: 3464 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 183 to 188 °C |
Titration Argentometric | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...